Anti-GPR3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-108EA 480 EUR
PRSI48-108
Anti-GPR3 Rabbit Polyclonal Antibody
Antikörper
GPR3 has been identified as a constitutively active lysophospholipid/lysosphingolipid receptor. GPR3 receptor is a link in communication between the somatic cells and oocyte of the ovarian follicle where GPR3 maintains meiotic arrest in mouse oocytes. Oocytes from Gpr3 knockout mice resume meiosis within antral follicles, independently of an increase in luteinizing hormone, and this phenotype Mice deficient in GPR3 exhibit premature ovarian failure due to early oocyte aging (Ledent et al. 2005).

GPR3 antibody can be used in ELISA, and immunohistochemistry starting at 3 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human, Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR