Anti-CCR10 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-106EA 480 EUR
PRSI48-106
Anti-CCR10 Rabbit Polyclonal Antibody
Antikörper
GPR21 is an Orphan-A GPCR with an unknown ligand.

GPR21 antibody can be used in immunohistochemistry starting at 20 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bat, Bovine, Dog, Gibbon, Gorilla, Human, Monkey, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR