Anti-CCR10 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-105EA 480 EUR
PRSI48-105
Anti-CCR10 Rabbit Polyclonal Antibody
Antikörper
GPR2/CCR10, a Chemokine Receptor, encodes a protein that most closely resembles the interleukin-8 receptor. The keratinocyte-produced chemokine CCL27 activates CCR10 and is involved in T cell-mediated skin inflammation.

CCR10 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CCR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR