Anti-GPR22 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI48-104EA 480 EUR
PRSI48-104
Anti-GPR22 Rabbit Polyclonal Antibody
Antikörper
GPR22 is an Orphan-A GPCR with no known ligand.

GPR22 antibody can be used in ELISA, and immunohistochemistry starting at 10 μg/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GPR22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bat, Bovine, Chicken, Xenopus, Gorilla, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Zebrafish
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR