Anti-FABP1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-098EA 619 EUR
PRSI48-098
Anti-FABP1 Mouse Monoclonal Antibody
Antikörper
FABP1 antibody can be used in Western Blot starting at 2 - 4 μg/mL, and immunohistochemistry starting at 5 μg/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FABP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR