Anti-NAPA Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-097EA 480 EUR
PRSI48-097
Anti-NAPA Mouse Monoclonal Antibody
Antikörper
SNAP antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.2, 0.1 mM PMSF, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NAPA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Dog, Hamster, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR