Anti-STX12 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-096EA 480 EUR
PRSI48-096
Anti-STX12 Mouse Monoclonal Antibody
Antikörper
Syntaxin 12 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.09% azide in 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: STX12
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Dog, Hamster, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR