Anti-SEPT5 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-095EA 480 EUR
PRSI48-095
Anti-SEPT5 Mouse Monoclonal Antibody
Antikörper
Septin 5 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, pH 7.2, 50% glycerol, 0.09% azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: SEPT5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR