Anti-B2M Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-094EA 480 EUR
PRSI48-094
Anti-B2M Mouse Monoclonal Antibody
Antikörper
Beta-2-Microglobulin antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, pH 7.4, 10 mM sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: B2M
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR