Anti-POMC Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-093EA 480 EUR
PRSI48-093
Anti-POMC Mouse Monoclonal Antibody
Antikörper
ACTH antibody can be used in Western Blot at 2 μg/mL, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation starting at 2 µg/mg lysate.

Protein A Column

PBS, pH 7.2, 0.09% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: POMC
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR