Anti-LMNA Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-091EA 480 EUR
PRSI48-091
Anti-LMNA Mouse Monoclonal Antibody
Lamin antibody can be used in ELISA starting at 1:6000 - 1:24000, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Tissue Culture Supernatant

0.2 M Tris/HCl, pH 7.4, 5-10% fetal calf serum.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LMNA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR