Anti-CFL1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-090EA 480 EUR
PRSI48-090
Anti-CFL1 Mouse Monoclonal Antibody
Antikörper
Cofilin antibody can be used in ELISA, Western Blot starting at 0.5 - 2 μg/mL, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation starting at 3 - 5 ng/1E6 cells

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CFL1
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR