Anti-APP Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-089EA 480 EUR
PRSI48-089
Anti-APP Mouse Monoclonal Antibody
Antikörper
APP antibody can be used in Western Blot starting at 1 μg/mL, and immunohistochemistry starting at 5 μg/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: APP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR