Anti-YWHAG Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-083EA 0 EUR
PRSI48-083
Anti-YWHAG Mouse Monoclonal Antibody
14-3-3 Gamma antibody can be used in immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: YWHAG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR