Anti-CTSL1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-082EA 480 EUR
PRSI48-082
Anti-CTSL1 Mouse Monoclonal Antibody
Antikörper
Cathepsin L antibody can be used in Western Blot starting at 1 μg/mL, and immunohistochemistry starting at 10 μg/mL.

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CTSL1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR