Anti-CYR61 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-079EA 0 EUR
PRSI48-079
Anti-CYR61 Mouse Monoclonal Antibody
CYR61 antibody can be used in Western Blot starting at 2.5 - 5 μg/mL, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYR61
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR