Anti-FOLH1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-070EA 480 EUR
PRSI48-070
Anti-FOLH1 Mouse Monoclonal Antibody
Antikörper
PSMA antibody can be used in Western Blot starting at 5 μg/mL, immunohistochemistry starting at 5 μg/mL, and immunoprecipitation.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOLH1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR