Anti-PSPH Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-055EA 480 EUR
PRSI48-055
Anti-PSPH Mouse Monoclonal Antibody
PSPH antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 20 - 40 μg/mL.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PSPH
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR