Anti-CEBPB Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-046EA 480 EUR
PRSI48-046
Anti-CEBPB Mouse Monoclonal Antibody
Antikörper
CEBP Beta antibody can be used in immunohistochemistry starting at 5 μg/mL.

PBS, pH 7.4, 0.1 % sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CEBPB
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR