Anti-LAIR1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-039EA 0 EUR
PRSI48-039
Anti-LAIR1 Mouse Monoclonal Antibody
LAIR-1 antibody can be used in immunohistochemistry starting at 2.5 μg/mL.

Protein G Column

PBS, pH 7.4, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LAIR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR