Anti-CYP26A1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-038EA 480 EUR
PRSI48-038
Anti-CYP26A1 Mouse Monoclonal Antibody
Antikörper
CYP26A1 antibody can be used in immunohistochemistry starting at 10 - 20 μg/mL.

Protein A/G Column

PBS, 0.08% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CYP26A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR