Anti-ENO2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-035EA 375 EUR
PRSI48-035
Anti-ENO2 Mouse Monoclonal Antibody
Antikörper
NSE antibody can be used in immunohistochemistry starting at 5 - 10 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ENO2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR