Anti-CFH Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-034EA 480 EUR
PRSI48-034
Anti-CFH Mouse Monoclonal Antibody
Antikörper
Factor H antibody can be used in immunohistochemistry starting at 2.5 - 5 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CFH
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR