Anti-UMOD Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-033EA 480 EUR
PRSI48-033
Anti-UMOD Mouse Monoclonal Antibody
Antikörper
Uromodulin antibody can be used in ELISA, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 10 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: UMOD
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR