Anti-CK13 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-031EA 480 EUR
PRSI48-031
Anti-CK13 Mouse Monoclonal Antibody
Antikörper
Cytoke antibody can be used in immunohistochemistry starting at 10 μg/mL, and flow cytometry starting at 1.5 - 2.5 μg/mL.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CK13
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR