Anti-PLP1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-029EA 480 EUR
PRSI48-029
Anti-PLP1 Mouse Monoclonal Antibody
Antikörper
PLP encodes the major protein components of compact CNS myelin and mutations in the PLP gene can lead to severe dysmyelinating disease.

PLP1 antibody can be used in ELISA starting at 1:400, immunohistochemistry starting at 5 μg/mL, and immunohistochemistry in frozen tissues starting at 1:50.

Protein A Column

PBS, 0.09% sodium azide

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Mammal
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR