Anti-TTR Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-026EA 480 EUR
PRSI48-026
Anti-TTR Mouse Monoclonal Antibody
Antikörper
Transthyretin antibody can be used in immunohistochemistry starting at 10 μg/mL.

Ammonium Sulfate Precipitation

HEPES with 0.15M NaCl, 0.01% BSA, 0.03% sodium azide, and 50% glycerol

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TTR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR