Anti-FGFR1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-018EA 604 EUR
PRSI48-018
Anti-FGFR1 Mouse Monoclonal Antibody
FGFR1 antibody can be used in immunohistochemistry starting at 1:10, and flow cytometry.

10 mM sodium HEPES, pH 7.5, 150 mM NaCl, 0.1 mg/mL BSA, 50% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FGFR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR