Anti-ERCC3 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-017EA 530 EUR
PRSI48-017
Anti-ERCC3 Mouse Monoclonal Antibody
XPB antibody can be used in immunohistochemistry starting at 10 μg/mL.

Protein G Column

0.1 M Tris-HCl, pH 7.5, before the addition of glycerol to 40%

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ERCC3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR