Anti-MET Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-016EA 480 EUR
PRSI48-016
Anti-MET Mouse Monoclonal Antibody
Antikörper
c-Met antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 μg/mL.

Purified

PBS, pH 7.4, 1% BSA, 0.1% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MET
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR