Anti-GFAP Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-014EA 480 EUR
PRSI48-014
Anti-GFAP Mouse Monoclonal Antibody
Antikörper
GFAP antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 15 μg/mL.

Protein G Column

PBS, pH 7.4, 0.2% BSA, 0.1% sodium azide, 40% glycerol.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GFAP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Human, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR