Anti-LRP1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-013EA 480 EUR
PRSI48-013
Anti-LRP1 Mouse Monoclonal Antibody
Antikörper
CD91 antibody can be used in immunohistochemistry starting at 2.5 μg/mL.

Protein A Column

0.05 M sodium phosphate; 0.1 M NaCl; 1 mM EDTA;, pH 6.6.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LRP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR