Anti-CR1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI48-010EA 480 EUR
PRSI48-010
Anti-CR1 Mouse Monoclonal Antibody
Antikörper
CD35 antibody can be used in ELISA starting at 1:000 - 1:1000, Western Blot starting at 1:100 - 1:500, and immunohistochemistry starting at 5 μg/mL.

Protein A Column

PBS, pH 7.4. Azide Free.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR