Anti-CD27 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: ProSci
PRSI48-006EA 480 EUR
PRSI48-006
Anti-CD27 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
CD27 antibody can be used in immunohistochemistry starting at 10 μg/mL.

Protein G Column

PBS, 0.02% sodium azide, EIA grade BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Tag
Antigen: CD27
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR