Anti-C4B Chicken Polyclonal Antibody

Lieferant: ProSci
PRSI48-001EA 480 EUR
PRSI48-001
Anti-C4B Chicken Polyclonal Antibody
Antikörper
C4B antibody can be used in immunohistochemistry starting at 20 μg/mL.

Purified

PBS, 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: 0
Antigen: C4B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR