Anti-KLK9 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI47-158EA 420 EUR
PRSI47-158
Anti-KLK9 Rabbit Polyclonal Antibody
Antikörper
PBS containing 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KLK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR