Anti-TDP2 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI47-141EA 420 EUR
PRSI47-141
Anti-TDP2 Mouse Monoclonal Antibody
Antikörper
The ski oncogene family (ski and sno) encodes transcriptional regulators that can affect oncogenic transformation and cellular differenti-ation. ski and sno function as co-activators or co-repressors, modulating transcription by inter-acting with transcription factor complexes. ski interacts with members of the Nuclear Factor I (NFI) family, the retinoic acid receptor, and the nuclear hormone co-regulators NCoR and NCoA62/skip.

Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TDP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken, Xenopus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR