Anti-THOC1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI47-132EA 553 EUR
PRSI47-132
Anti-THOC1 Rabbit Polyclonal Antibody
Antikörper
Prostein is a novel prostate-specific protein expressed in normal and malignant prostate tissues. This protein appears tobe a type IIIa plasma membrane protein with a cleavable signal peptide and 11 transmembrane-spanning regions. Prostein expression is androgen responsive since treatment of LNCaP cells with androgen up-regulates prostein message and protein expression levels. Thus, prostein is a prostate-specific marker whose detection and quantitationmay be useful in diagnosing and monitoring prostate cancer.

PBS containing 0.02% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: THOC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR