Anti-LIG3 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI47-131EA 420 EUR
PRSI47-131
Anti-LIG3 Rabbit Polyclonal Antibody
Antikörper
Epithelium-specific gene regulation plays acritical role during embryogenesis, and the epithelial cell lineage is the first differentiated cell type to appear after fertilization. Until recently, very few epithelial cell-restricted transcription factors had been described. Members of the ets transcription factor/oncogene family have been implicated in human carcinogenesis following the discovery of several distinctchromosomal translocations involving members of the ets family in different types of cancer. ets factors play key roles intranscriptional control of stringently regulated genes, such as genes involved in tissue development, differentiation,angiogenesis, cell cycle control and cellproliferation. ESE-1 is the first member ofthe ets family that was found to be exclusively expressed and inducible inepithelial cells.

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LIG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR