Anti-LIG1 Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI47-130EA 455 EUR
PRSI47-130
Anti-LIG1 Mouse Monoclonal Antibody
Antikörper
Thymidine kinase (TK) belongs to a group of enzymes such as dihydrofolate reductase, thymidylate synthase, and DNA polymerase that are involved in DNA synthesis and precursor production. High levels of these enzymes are present in proliferating cells, and low levels are found in resting cells, due to multiple regulatory mechanisms that ensure exclusive expression of these enzymes in replicating cells. TK is responsible for catalyzing the phosphorylation of thymidine, which functions as a part of the pyrimidine salvage pathway involved in DNA synthesis. The activities of enzymes such as TK are essential for the activation of several chemotherapeutically important nucleoside analogues that are administered as prodrugs.

Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LIG1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR