Anti-MRE11A Mouse Monoclonal Antibody

Lieferant: ProSci
PRSI47-127EA 505 EUR
PRSI47-127
Anti-MRE11A Mouse Monoclonal Antibody
Antikörper
Protein G-purified

PBS

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MRE11A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR