Anti-NBN Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI47-108EA 612 EUR
PRSI47-108
Anti-NBN Rabbit Polyclonal Antibody
Antikörper
1 mg/ml BSA, 0.05% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: NBN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR