Anti-RTEL1 Rabbit Polyclonal Antibody

Lieferant: ProSci
PRSI47-106EA 480 EUR
PRSI47-106
Anti-RTEL1 Rabbit Polyclonal Antibody
Antikörper
1 mg/ml BSA, 0.05% sodium azide.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RTEL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR