Anti-F2R Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-994EA 420 EUR
PRSI46-994
Anti-F2R Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 48 kDa band observed in HeLa lysate (calculated MW of 47.4 kDa according to NP_001983.2). Recommended concentration: 0.5-2 μg/mL.

PAR1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PAR1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: F2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR