Anti-CAV3 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-992EA 420 EUR
PRSI46-992
Anti-CAV3 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:4000. Western Blot: Approximately 23 kDa band observed in human, mouse and rat heart lysates (calculated MW of 17.3 kDa according to NP_203123.1). The observed molecular weight corresponds to earlier findings with different antibodies from other commercial sources. Recommended concentration: 0.3-1 μg/mL.

Caveolin-3 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Caveolin-3 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: CAV3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR