Anti-PSPH Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-991EA 420 EUR
PRSI46-991
Anti-PSPH Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 26 kDa band observed in human hippocampus and in mouse brain lysates (calculated MW of 25.0 kDa according to human NP_004568.2 and 25.1 kDa according to mouse NP_598661.1). Recommended concentration: 0.05-1 μg/mL.

PSPH antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PSpH antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PSPH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR