Anti-DLL1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-984EA 420 EUR
PRSI46-984
Anti-DLL1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Approximately 75 kDa band observed in mouse heart and rat heart lysates (calculated MW of 78.1 kDa according to human NP_005609.3, 78.5 kDa according to mouse NP_031891.2 and 77.4 kDa according to rat NP_114452.1). Recommended concentration: 0.3-1 μg/mL.

DLL1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

DLL1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DLL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR