Anti-GCKR Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-981EA 553 EUR
PRSI46-981
Anti-GCKR Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 70 kDa band observed in mouse and rat liver lysates (calculated MW of 68.7 kDa according to human NP_001477.1 and of 68.9 kDa according to rat NP_037252.1). Recommended concentration: 0.1-0.3 μg/mL.

GCKR antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

GCKR antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GCKR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR