Anti-KALRN Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-975EA 345 EUR
PRSI46-975
Anti-KALRN Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:64000. Approximately 49 kDa band in human, mouse and rat brain lysates after 0.3 μg/mL antibody staining.The 49 kDa band was successfully blocked by incubation with the immunizing peptide.

KALRN antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

KALRN antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: KALRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR