Anti-PLEKHB1 Goat Polyclonal Antibody

Lieferant: ProSci
PRSI46-974EA 420 EUR
PRSI46-974
Anti-PLEKHB1 Goat Polyclonal Antibody
Antikörper
ELISA: Antibody detection limit dilution 1:16000. Approximately 25 kDa band observed in rat brain lysates (calculated MW of 25.2 kDa according to rat NP_742030.1 (Evectin-1) and of 27.2 kDa according to human NP_067023.1). Recommended concentration: 1-3 μg/mL.

PHR1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

pH R1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLEKHB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR